Tisk…
Cole-Parmer® FJT-200 Series Flocculation Jar Testers, Stuart
Dodavatel: Cole Parmer
99551-00 99551-05
STUA99551-00EA 83930 CZK
STUA99551-00 STUA99551-05
Cole-Parmer® FJT-200 Series Flocculation Jar Testers, Stuart
Flokulátory
Get repeatable conditions between samples.
  • Easily view paddle speed on digital display
  • Save time with two pre-set programs
  • Built-in illumination and dark background for easy viewing of samples
  • Max Stirring Volume per position: 1 L
Achieve repeatable conditions between samples and from run to run with these flocculation jar testers. Speed control with electronic feedback accurately maintains the rotational speed of the stainless-steel paddles. Two preset programs allow routine speed and time parameters to be entered, stored in memory and recalled orMore Product Information