Tisk…

Zinc D-gluconate ≥97.0% USP

Dodavatel: Spectrum Chemical

Synonyms: Zinc gluconate

Z1077-12KG Z1077-50KGBL Z1077-2.5KG
SPCMZ1077-12KGEA 80890 CZK
SPCMZ1077-12KG SPCMZ1077-50KGBL SPCMZ1077-2.5KG
Zinc D-gluconate ≥97.0% USP
Zinc D-gluconate

ZINC GLUCONATE, USP is a zinc salt of gluconic acid can also be manufactured by fermenting glucose. It comes as a white powder and is used in cold treating lozenges and as one of the more effective ways of delivering zinc as a dietary supplement. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: C₁₂H₂₂O₁₄Zn
MDL Number: MFCD00868110
CAS číslo: 4468-02-4
EINECS: 224-736-9

Objednat

Výsledky specifikačního testu

Assay (Anhydrous Basis) 97.0 - 102.0 %
pH of a 1 in 100 Solution 5.5 - 7.5
Water max. 11.6 %
Chloride (Cl) max. 0.05 %
Sulfate max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR