Tisk…

Sodium stearyl fumarate 99.0-101.5% (by anhydrous basis) NF

Dodavatel: Spectrum Chemical

S1448-25KGBL S1448-500GM S1448-25GM
SPCMS1448-25KGBLEA 0 CZK
SPCMS1448-25KGBL SPCMS1448-500GM SPCMS1448-25GM
Sodium stearyl fumarate 99.0-101.5% (by anhydrous basis) NF
Sodium stearyl fumarate

Sodium stearyl fumarate, NF is a water-soluble tablet lubicant.


Manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: C₂₂H₄₀O₄
MDL Number: MFCD00236640
CAS číslo: 4070-80-8

Objednat

Výsledky specifikačního testu

Assay (Anhydrous Basis) 99.0 - 101.5%
Saponification Value 142.2 - 146.0
Water Determination Max. 5.0%
Lead (Pb) Max. 10 ppm
Heavy Metals Max. 20 ppm
Sodium Stearyl Maleate And Stearyl Alcohol To Pass Test
Residual Solvents To Pass Test

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR