Tisk…

D-Panthenol ≥98% USP

Dodavatel: Spectrum Chemical

P1831-45KGBL P1831-1KG
SPCMP1831-45KGBLEA 611530 CZK
SPCMP1831-45KGBL SPCMP1831-1KG
D-Panthenol ≥98% USP
D-Panthenol

Dexpanthenol, USP is the alcohol analog of pantothenic acid, and is thus a provitamin of B5.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: C₉H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00065006
CAS číslo: 81-13-0
EINECS: 201-327-3

Objednat

Výsledky specifikačního testu

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR