Tisk…

Potassium hydroxide, pellets FCC

Dodavatel: Spectrum Chemical

P1316-2.5KG P1316-500GM P1316-12KG P1316-50KGBL
SPCMP1316-2.5KGEA 17380 CZK
SPCMP1316-2.5KG SPCMP1316-500GM SPCMP1316-12KG SPCMP1316-50KGBL
Potassium hydroxide, pellets FCC
Hydroxid draselný

Potassium Hydroxide, Pellets, FCC is used as a rinse or chemical peel for fruits and vegetables, a stabiliser to prolong shelf-life, and as a thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Skull and crossbones
UN1813

Vzorec: KOH
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS číslo: 1310-58-3
UN: 1813
ADR: 8,II

Objednat

Výsledky specifikačního testu

Assay (Total Alkali As KOH) 85.0 - 100.5%
Carbonate (As K₂CO₃) max. 3.5 %
Insoluble Substances To Pass Test
Lead (Pb) max. 2 mg/kg
Mercury max. 0.1 MG/


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR