Tisk…

Poloxamer 407 (Pluronic® F-127) NF

Dodavatel: Spectrum Chemical

P1166-2.5KG P1166-45KGBL P1166-500GM P1166-12KG P1166-25KGBL
SPCMP1166-2.5KGEA 19860 CZK
SPCMP1166-2.5KG SPCMP1166-45KGBL SPCMP1166-500GM SPCMP1166-12KG SPCMP1166-25KGBL
Poloxamer 407 (Pluronic® F-127) NF
Poloxamer 407 (Pluronic® F-127)

Poloxamer 407, NF is a national formulary approved surfactant that finds utility because of its amphiphilic structure.


  • Amphphilic structure
  • Increases miscibility
  • Controls thickness of personal care products


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Vzorec: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number: MFCD00082049
CAS číslo: 9003-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR