Tisk…

Itraconazole, micronised USP

Dodavatel: Spectrum Chemical

Synonyms: Oriconazole

I2662-100GM I2662-1KGBL I2662-25KGBL I2662-5KGBL
SPCMI2662-100GMEA 91920 CZK
SPCMI2662-100GM SPCMI2662-1KGBL SPCMI2662-25KGBL SPCMI2662-5KGBL
Itraconazole, micronised USP
Itraconazole

Itraconazole, Micronised, USP is a drug used for the treatment of fungal infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: C₃₅H₃₈Cl₂N₈O₄
MW: 705.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00870168
CAS číslo: 84625-61-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR