Tisk…

Imidazolidinyl urea NF

Dodavatel: Spectrum Chemical

I1007-12KG I1007-50KGBL
SPCMI1007-12KGEA 119180 CZK
SPCMI1007-12KG SPCMI1007-50KGBL
Imidazolidinyl urea NF
Imidazolidinyl urea

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Vzorec: C₁₁H₁₆N₈O₈
Storage Temperature: Ambient
MDL Number: MFCD00221482
CAS číslo: 39236-46-9

Objednat

Výsledky specifikačního testu

Assay (As Nitrogen) 26.0 - 28.0%
Color and Clarity of Solution To Pass Test
pH of a 1% Solution 6.0 - 7.5
Loss On Drying max. 3.0%
Residue On Ignition max. 3.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR