Tisk…

Carbomer 934P resin, powder NF

Dodavatel: Spectrum Chemical

Synonyms: Carbopol 934P

CA184-100GM
SPCMCA184-100GMEA 7510 CZK
SPCMCA184-100GM
Carbomer 934P resin, powder NF
Carbomer 934P

Carbomer 934P, Resin, NF polymers can provide bioadhesion and good binding characteristics that provide highly effective controlled-release properties.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Storage Temperature: Ambient
MDL Number: MFCD00084394
CAS číslo: 9062-04-8

Objednat

Výsledky specifikačního testu

Carboxylic Acid Groups (Dried Basis) 56.0 - 68.0%
Viscosity of a 0.5% Solution 29,400-39,400 mPa·s
Loss On Drying max. 2.0%
Heavy Metals max. 20 µg/g
Benzene max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR