Tisk…

Calcium hydroxide 95,0, powder FCC

Dodavatel: Spectrum Chemical

Synonyms: Slaked lime, Calcium dihydroxide

C1116-2.5KG
SPCMC1116-2.5KGEA 10850 CZK
SPCMC1116-2.5KG
Calcium hydroxide 95,0, powder FCC
Hydroxid vápenatý

Calcium Hydroxide, Powder, FCC is used extensively in the food industry to clarify solutions, as a digestion aid, baking soda substitute and as a nutritional supplement because of its low toxicity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark.

Vzorec: Ca(OH)₂
MW: 74.09
Bod tání: 550
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS číslo: 1305-62-0
UN: 3262
ADR: 8 III
Merck Index: 13,01675

Objednat

Výsledky specifikačního testu

Assay [Ca(OH)₂] 95.0 - 100.5%
Acid-Insoluble Substances max. 0.5 %
Arsenic (As) max. 3 mg/kg
Carbonate To Pass Test
Fluoride max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR