Tisk…

D-(+)-Biotin ≥97.5%, powder FCC

Dodavatel: Spectrum Chemical

BI115-500GMBL BI115-1GM BI115-100GMBL BI115-25GM
SPCMBI115-500GMBLEA 346820 CZK
SPCMBI115-500GMBL SPCMBI115-1GM SPCMBI115-100GMBL SPCMBI115-25GM
D-(+)-Biotin ≥97.5%, powder FCC
D(+)-Biotin

Biotin, Powder, FCC is often recommended as a dietary supplement for strenghening hair and nails. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: C₁₀H₁₆N₂O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS číslo: 58-85-5

Objednat

Výsledky specifikačního testu

Assay (C₁₀H₁₆N₂O₃S) 97.5 - 100.5%
Melting Range 229 - 232°C
Optical Rotation +89 to +93
Lead (Pb) max. 2 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR