Tisk…

p-Anisaldehyde 98, reagent

Dodavatel: Spectrum Chemical

Synonyms: 4-Methoxybenzaldehyde

AN122-10LTBL
SPCMAN122-10LTBLEA 0 CZK
SPCMAN122-10LTBL
p-Anisaldehyde 98, reagent
p-Anizaldehyd

p-Anisaldehyde, Reagent, also known as 4-Anisaldehyde, is considered a useful stain in thin layer chromatography. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Vzorec: C₈H₈O₂
MW: 136.15
Bod varu: 140 °C (1013 hPa)
Bod tání: 0…2 °C
Hustota: 1,119
Bod vzplanutí: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003385
CAS číslo: 123-11-5
EINECS: 204-602-6
Merck Index: 13,00666

Objednat

Výsledky specifikačního testu

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR