Tisk…

Hydrogenfosforečnan diamonný ≥96.0% FCC

Dodavatel: Spectrum Chemical

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

AM184-12KG AM184-500GM
SPCMAM184-12KGEA 61280 CZK
SPCMAM184-12KG SPCMAM184-500GM
Hydrogenfosforečnan diamonný ≥96.0% FCC
Hydrogenfosforečnan diamonný

Ammonium Phosphate Dibasic, FCC is a food ingredient used as an acidity regulator and raising agent. It is listed on the FDA's Generally Recognized As Safe (GRAS) list of substances. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Vzorec: (NH₄)₂HPO₄
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS číslo: 7783-28-0

Objednat

Výsledky specifikačního testu

Assay [(NH₄)₂HPO₄] 96.0 - 102.0 %
Arsenic (As) max. 3 mg/kg
Fluoride max. 10 mg/kg
Lead (Pb) max. 4 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR