Tisk…
BIOS A and BIOS 16 Centrifuges with GreenCool Technology
Dodavatel: SORVALL
75008612 75008614 75008617 75008620 75008621 75008622
SORV75008612EA 0 CZK
SORV75008612 SORV75008614 SORV75008617 SORV75008620 SORV75008621 SORV75008622
BIOS A and BIOS 16 Centrifuges with GreenCool Technology
Centrifugy Centrifugy na zemi stojící
Thermo Scientific™ BIOS A and BIOS 16 Centrifuges with GreenCool Technology bring enhanced sustainability to bioprocessing facilities and labs performing high-throughput applications. Featuring a natural refrigerant (CO₂) cooling system, they offer reduced energy consumption, rapid cool-down times while their lighter weight and reduced noise levels help improve the laboratory environment.
  • Global Warming Potential (GWP) of 1, up to 15% lower energy consumption
  • Large capacity of up to 16 L
  • Auto-ID instant rotor identification automatically and instantly
More Product Information