Tisk…
Qualitix® Pipette Tips
Dodavatel: Socorex Isba
302.0010GR 312.05B 307.1000B 307.0200B 308.0350R 312.05R 327.0200B 308.0350B 312.02 312.10 3020.0010B 3190.1000R 3020.0010PR 3020.0010PRS 3020.0010R 3070.0200B 3070.0200PR 3070.0200PRS 3070.0200R 3070.0200RS 3070.0200T 3070.1000B 3070.1000PR 3070.1000PRS 3070.1000R 3070.1000RS 3070.1000T 3080.0200GR 3090.0010B 3090.0010PR 3090.0010PRS 3090.0010R 3090.0010T 3090.1000B 3090.1000R 3190.1000B 3290.0200B 3290.0200R 3290.0200RS
SOCO302.0010GREA 3490 CZK
SOCO302.0010GR 613-0575 SOCO307.1000B SOCO307.0200B SOCO308.0350R 613-0573 SOCO327.0200B SOCO308.0350B SOCO312.02 613-5824 SOCO3020.0010B SOCO3190.1000R SOCO3020.0010PR SOCO3020.0010PRS SOCO3020.0010R SOCO3070.0200B SOCO3070.0200PR SOCO3070.0200PRS SOCO3070.0200R SOCO3070.0200RS SOCO3070.0200T SOCO3070.1000B SOCO3070.1000PR SOCO3070.1000PRS SOCO3070.1000R SOCO3070.1000RS SOCO3070.1000T SOCO3080.0200GR SOCO3090.0010B SOCO3090.0010PR SOCO3090.0010PRS SOCO3090.0010R SOCO3090.0010T SOCO3090.1000B SOCO3090.1000R SOCO3190.1000B SOCO3290.0200B SOCO3290.0200R SOCO3290.0200RS
Qualitix® Pipette Tips
Špičky pipetovací
These tips are made of 100% pure virgin, medical grade PP, fitting not only Socorex® but all major micropipette brands.
  • Superior quality, autoclavable
  • Lot by lot consistency
  • Certified free of nuclease, endotoxin and ATP
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with other micropipette brands
Beveled tips: Special shape eliminates liquid drop sticking at tip end.
Fine pointed tips: Have a thin, soft end enabling smooth liquid deposit.
Low binding tips are made with a silicone-free resin reducing surface adhesion. TheyMore Product Information