Tisk…
Random hexamer primer
Dodavatel: Thermo Fisher Scientific
SO142
SO142EA 1850 CZK
SO142
Random hexamer primer
Činidla pro nukleové kyseliny Primers and Probes
Random hexamer primer is a mixture of single-stranded random hexanucleotides with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Random hexamer primer has been functionally tested in first strand cDNA synthesis.