Tisk…
Zarate Trousers with ARC Protection
Dodavatel: SIOEN
11VA2PFAB9854R 11VA2PFAH4652L 11VA2PFAH4650R 11VA2PFAH4650L 11VA2PFAH4648R 11VA2PFAH4648L 11VA2PFAH4646R 11VA2PFAH4646L 11VA2PFAB9846L 11VA2PFAH4644R 11VA2PFAB9864R 11VA2PFAB9862R 11VA2PFAB9860R 11VA2PFAB9858R 11VA2PFAB9856L 11VA2PFAB9854L 11VA2PFAB9852R 11VA2PFAB9852L 11VA2PFAB9850R 11VA2PFAB9850L 11VA2PFAB9848R 11VA2PFAB9848L 11VA2PFAB9846R 11VA2PFAB9844R 11VA2PFAB9856R 11VA2PFAH4652R 11VA2PFAM4464R 11VA2PFAM4462R 11VA2PFAM4460R 11VA2PFAH4656R 11VA2PFAM4456R 11VA2PFAM4456L 11VA2PFAM4458R 11VA2PFAH4660R 11VA2PFAH4654L 11VA2PFAH4658R 11VA2PFAH4656L 11VA2PFAH4654R 11VA2PFAM4454R 11VA2PFAM4454L 11VA2PFAM4452R 11VA2PFAM4452L 11VA2PFAM4450R 11VA2PFAM4450L 11VA2PFAM4448R 11VA2PFAM4448L 11VA2PFAM4446R 11VA2PFAM4446L 11VA2PFAM4444R 11VA2PFAH4664R 11VA2PFAH4662R 11VA2PFAB9848S 11VA2PFAB9850S 11VA2PFAB9852S 11VA2PFAB9854S 11VA2PFAB9856S 11VA2PFAB9858S 11VA2PFAH4648S 11VA2PFAH4650S 11VA2PFAH4652S 11VA2PFAH4654S 11VA2PFAH4656S 11VA2PFAH4658S 11VA2PFAM4448S 11VA2PFAM4450S 11VA2PFAM4452S 11VA2PFAM4454S 11VA2PFAM4456S 11VA2PFAM4458S
SIOE11VA2PFAB9854REA 3130 CZK
SIOE11VA2PFAB9854R SIOE11VA2PFAH4652L SIOE11VA2PFAH4650R SIOE11VA2PFAH4650L SIOE11VA2PFAH4648R SIOE11VA2PFAH4648L SIOE11VA2PFAH4646R SIOE11VA2PFAH4646L SIOE11VA2PFAB9846L SIOE11VA2PFAH4644R SIOE11VA2PFAB9864R SIOE11VA2PFAB9862R SIOE11VA2PFAB9860R SIOE11VA2PFAB9858R SIOE11VA2PFAB9856L SIOE11VA2PFAB9854L SIOE11VA2PFAB9852R SIOE11VA2PFAB9852L SIOE11VA2PFAB9850R SIOE11VA2PFAB9850L SIOE11VA2PFAB9848R SIOE11VA2PFAB9848L SIOE11VA2PFAB9846R SIOE11VA2PFAB9844R SIOE11VA2PFAB9856R SIOE11VA2PFAH4652R SIOE11VA2PFAM4464R SIOE11VA2PFAM4462R SIOE11VA2PFAM4460R SIOE11VA2PFAH4656R SIOE11VA2PFAM4456R SIOE11VA2PFAM4456L SIOE11VA2PFAM4458R SIOE11VA2PFAH4660R SIOE11VA2PFAH4654L SIOE11VA2PFAH4658R SIOE11VA2PFAH4656L SIOE11VA2PFAH4654R SIOE11VA2PFAM4454R SIOE11VA2PFAM4454L SIOE11VA2PFAM4452R SIOE11VA2PFAM4452L SIOE11VA2PFAM4450R SIOE11VA2PFAM4450L SIOE11VA2PFAM4448R SIOE11VA2PFAM4448L SIOE11VA2PFAM4446R SIOE11VA2PFAM4446L SIOE11VA2PFAM4444R SIOE11VA2PFAH4664R SIOE11VA2PFAH4662R SIOE11VA2PFAB9848S SIOE11VA2PFAB9850S SIOE11VA2PFAB9852S SIOE11VA2PFAB9854S SIOE11VA2PFAB9856S SIOE11VA2PFAB9858S SIOE11VA2PFAH4648S SIOE11VA2PFAH4650S SIOE11VA2PFAH4652S SIOE11VA2PFAH4654S SIOE11VA2PFAH4656S SIOE11VA2PFAH4658S SIOE11VA2PFAM4448S SIOE11VA2PFAM4450S SIOE11VA2PFAM4452S SIOE11VA2PFAM4454S SIOE11VA2PFAM4456S SIOE11VA2PFAM4458S
Zarate Trousers with ARC Protection
Pracovní oděvy
Multifunctional and anti-static ARC trousers that provide excellent protection against arcs, heat, and metal splashes.
  • Closure by 1 hidden button/front fly with zip closure
  • 2 inset pockets with flap
  • 1 back pocket with flap closed by concealed press studs
  • 1 patched thigh pocket with flap closed by hidden press studs
  • 1 ruler pocket with flap closed by concealed press studs
  • Elasticated waist
  • Knee pockets
  • Waistband with belt loops
The trousers are made of a breathable and non-allergenic fabric and are very practical with two inset pockets with flap, oneMore Product Information