Tisk…
Screw caps for microcentrifuge tubes
Dodavatel: Simport Scientific
T366NLS T366BLS T366GLS T366RLS T366WLS T366YLS T366AMLS T366NLSL T366BLSL T366GLSL T366RLSL T366WLSL T366YLSL T366AMLSL
SIMPT366NLSEA 1180 CZK
SIMPT366NLS SIMPT366BLS SIMPT366GLS SIMPT366RLS SIMPT366WLS SIMPT366YLS SIMPT366AMLS SIMPT366NLSL SIMPT366BLSL SIMPT366GLSL SIMPT366RLSL SIMPT366WLSL SIMPT366YLSL SIMPT366AMLSL
Screw caps for microcentrifuge tubes
Uzávěry Víčka na zkumavky
Flat top closures with or without loop are designed with cavity for 2D and CapInsert™.
  • Non-sterile