Tisk…
Microcentrifuge tubes, ClikLok™ and MacrewTube™
Dodavatel: Simport Scientific
T330-76N T330-75N T330-75B T330-75G T330-75R T330-75Y T330-75AM T330-75NS T330-75AMS T330-76B T330-76G T330-76R T330-76Y T330-76AM T330-76NS T330-76AMS T365-5TN T365-5TAM T365-5NLSS
SIMPT330-76NEA 2180 CZK
SIMPT330-76N SIMPT330-75N SIMPT330-75B SIMPT330-75G SIMPT330-75R SIMPT330-75Y SIMPT330-75AM SIMPT330-75NS SIMPT330-75AMS SIMPT330-76B SIMPT330-76G SIMPT330-76R SIMPT330-76Y SIMPT330-76AM SIMPT330-76NS SIMPT330-76AMS SIMPT365-5TN SIMPT365-5TAM SIMPT365-5NLSS
Microcentrifuge tubes, ClikLok™ and MacrewTube™
Zkumavky Mikrozkumavky centrifugační
These tubes are made of high-quality, transparent polypropylene without the use of slip agents, plasticisers, mold release agents and biocide substances.
  • RNase-, DNase-, Pyrogen- and DNA-free
  • Writing surface on flat cap and side wall for quick sample identification
  • Precise lid sealing for minimal evaporation rate during long-term storage
  • Graduated every 0,5 ml
  • Exceptional centrifugation stability of up to 25000×g
  • Withstands temperatures from +121 to –196 °C
The ClikLok™ and MacrewTube™ tubes have been designed for the simple and safe processing ofMore Product Information