Tisk…
Cluster tubes, 2 ml, and storage racks, BioTube™ T105
Dodavatel: Simport Scientific
T105-50 T105-51 T105-20LST T105-20
SIMPT105-50EA 6250 CZK
SIMPT105-50 SIMPT105-51 SIMPT105-20LST SIMPT105-20
Cluster tubes, 2 ml, and storage racks, BioTube™ T105
Zkumavky Mikrozkumavky
These racks contain 96 removable PP square tubes in a 8×12 configuration, each having a 2,1 ml capacity (2 ml when capped). Tubes are also available separately.
  • Racks are stackable to save on storage space
  • With PVC cover for full protection of tube contents
  • Square tubes feature a round bottom to facilitate emptying
  • Certified RNase, DNase, pyrogen- and DNA-free
  • Conforms to SBS standards
  • Available in sterile and non sterile versions
Compatible with most robotic workstations, the PP storage racks can be used with most cell harvesters and leading 8- and 12More Product Information