Tisk…
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Dodavatel: Simport Scientific
M525-2T M525-3T M525-4T M525-5T M525-6T M525-7T M525-8T M525-9T M525-10T M525-11T M525-12T
SIMPM525-2TEA 6100 CZK
SIMPM525-2T SIMPM525-3T SIMPM525-4T SIMPM525-5T SIMPM525-6T SIMPM525-7T SIMPM525-8T SIMPM525-9T SIMPM525-10T SIMPM525-11T SIMPM525-12T
Tissue processing/embedding cassettes in QuickLoad™ stacks, Unisette™, M525T
Kazety pro zalévání vzorků Kazety tkáňové
These cassettes are moulded from acetal and keep specimens safely submerged in liquid, and are resistant to the chemical action of most histological solvents.
  • Anterior writing area at a 45° angle
  • Taped cassettes to be used with Leica ink jet printers
  • Easy to remove disposable cover
Unisette™ histology cassette comes with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
The efficient flow-through slots maximise fluid exchange and ensureMore Product Information