Tisk…
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518
Dodavatel: Simport Scientific
M518-10T M518-11T M518-12T M518-2T M518-3T M518-4T M518-5T M518-6T M518-7T M518-8T M518-9T
SIMPM518-10TEA 13970 CZK
SIMPM518-10T SIMPM518-11T SIMPM518-12T SIMPM518-2T SIMPM518-3T SIMPM518-4T SIMPM518-5T SIMPM518-6T SIMPM518-7T SIMPM518-8T SIMPM518-9T
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518
Kazety pro zalévání vzorků Kazety bioptické
Secure specimens during processing with Swingsette™ Biopsy processing/embedding cassette, in Quickload™ stacks.
  • Anterior writing area, 45°
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
  • Large tab for convenient and easy opening
They keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximise fluid exchange and ensure proper drainage.
These cassettes are especiallyMore Product Information