Tisk…
Colorimeter, spectral, X-Rite RM200QC
Dodavatel: Industrial Physics
VF0620
SIMEVF0620EA 96820 CZK
SIMEVF0620
Colorimeter, spectral, X-Rite RM200QC
Kolorimetry Kolorimetry reflexní
The X-Rite RM200QC imaging spectrocolorimeter bridges the gap between colour appearance and material colour from incoming material batches to outgoing product shipments. It is designed to provide stable colour comparisons for materials and products wherever colour control is important.
  • Build a complete colour program
  • Better working agreement
  • Increased quality control
  • Quick responsive tools
Operating conditions: 0 to 40 °C; 20 to 80 % RH.