Tisk…
Indirect impact tester
Dodavatel: Industrial Physics
SP1880
SIMESP1880EA 69690 CZK
SIMESP1880
Indirect impact tester
Testery nátěrů
Impact testers used to determine the impact resistivity and flexibility of materials.
  • Spirit level built-in for correct positioning
  • Dual scale instrument equipped with a special guidance
  • Distance between each impact is always according to the standards
Certifikace: ISO 6272-2 and ASTM D2794