Tisk…
Sound level meter, digital
Dodavatel: Industrial Physics
LU0115
SIMELU0115EA 6870 CZK
SIMELU0115
Sound level meter, digital
Měřiče zvuku
Easy to use digital sound level meter with wide dynamic range, 30 to 130 dB in the frequency range 31,5 Hz to 8 kHz
  • With electret condenser microphone
  • Hold function for maximum values
  • Illuminated digital display with 4 digits, resolution 0,1 dB
It has different frequency weighting A and C to respond to human sense or machine monitoring purposes. For different sound patterns, it also supplies Fast and Slow time constant setting.
Informace o dodávce: Supplied with battery, carrying case and manual.