Tisk…

Surface profile readers, PosiTector® RTR, DeFelsko®

Dodavatel: Industrial Physics
LD5430 LD5431 LD5432 LD5433 LD5434
SIMELD5430EA 29880 CZK
SIMELD5430 SIMELD5431 SIMELD5432 SIMELD5433 SIMELD5434
Surface profile readers, PosiTector® RTR, DeFelsko®
Testery nátěrů
These digital spring micrometer measures and records peak to valley surface profile height using replica tape reader and it provides simple, durable, accurate and versatile reading.

  • Solvent, acid, oil, water and dust resistant – weatherproof
  • Continually displays average, standard deviation, min/max and number of readings
  • Storage of 250 readings (Standard) or 100 000 readings (Advanced)
  • Includes PosiSoft suite of solutions for analyzing and reporting data
  • Transfer data via USB to a PC/Mac and also via Bluetooth wireless technology and WiFi technology in
More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR