Tisk…
Reference guide and photographs for steel surfaces, SSPC-VIS 1
Dodavatel: Industrial Physics
LD3055
SIMELD3055EA 11490 CZK
SIMELD3055
Reference guide and photographs for steel surfaces, SSPC-VIS 1
Knihy
The SSPC is using visual reference, that features over 50 full colour pictures of (coated and uncoated) rusted steel surfaces, before and after dry abrasive blast cleaning.
Cleanliness requirements for conditions depicted are defined by the SSPC surface preparation specifications for white metal (SP 5), near-white (SP 10), commercial (SP 6), industrial (SP 14), and brush-off (SP 7) blast finishes. Appendix photographs show variations in white metal surfaces caused by different metallic and non-metallic abrasives, profile depth, angle of view, and lighting. Guides available in both EnglishMore Product Information