Tisk…
Rust Grade Book, ISO 8501-1
Dodavatel: Industrial Physics
LD3020
SIMELD3020EA 20010 CZK
SIMELD3020
Rust Grade Book, ISO 8501-1
Knihy
Pictorial standard book with rust grades and preparation grades of uncoated steel substrates and of steel substrates after overall removal of previous coatings. Also called 'Swedish Blast Degrees' or Sa-standards). The original visual standard. It shows the degree of cleanliness of four different levels of rusted steel cleaned by grit blasting, hand/power tool and flame cleaning.