Tisk…
Temperature loggers, CurveX-3 basic oven logger
Dodavatel: Industrial Physics
CX3005 CX3010
SIMECX3005EA 39310 CZK
SIMECX3005 SIMECX3010
Temperature loggers, CurveX-3 basic oven logger
Záznamníky dat (Dataloggery)
CurveX 3 basic is a 4-channel oven recorder specially designed for powder coating lines.
  • Operate through only three buttons
  • Factory calibrated for immediate use
  • Memory for ten different batches, automatically overwrites the oldest results
  • Programmable 'paint type' memory for immediate 'pass/fail' result
  • Flat design, only 16 mm in Ø, for use in low clearance ovens
  • Built in a sturdy machined aluminium case that fulfills the basic needs for quality control in powder coating applications
It features durable and impact resistant housing, USB connectors,More Product Information