Tisk…

Diethyl disulphide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ300

W409301-SAMPLE W409301-1KG
SIALW409301-SAMPLEEA 1750 CZK
SIALW409301-SAMPLE SIALW409301-1KG
Diethyl disulphide, Sigma-Aldrich®
Diethyl disulphide
Vzorec: C₄H₁₀S₂
Bod varu: 152 °C (1013 hPa)
Hustota: 0,984 g/cm³ (20 °C)
MDL Number: MFCD00009266
CAS číslo: 110-81-6
EINECS: 203-805-7
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR