Tisk…

trans-2-Heptenal, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W316504-1KG-K W316504-200G-K
SIALW316504-1KG-KEA 25230 CZK
SIALW316504-1KG-K SIALW316504-200G-K
trans-2-Heptenal, Sigma-Aldrich®
trans-2-Heptenal
Vzorec: C₇H₁₂O
Bod varu: 90 °C (1013 hPa)
Hustota: 0,8575 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007010
CAS číslo: 18829-55-5
EINECS: 242-608-0
UN: 1988
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR