Tisk…

(±)-α-Terpineol, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ300

W304506-10KG-K W304506-1KG-K W304506-25KG-K
SIALW304506-10KG-KEA 8150 CZK
SIALW304506-10KG-K SIALW304506-1KG-K SIALW304506-25KG-K
(±)-α-Terpineol, Sigma-Aldrich®
(±)-α-Terpineol
Vzorec: C₁₀H₁₈O
MDL Number: MFCD00001557
CAS číslo: 98-55-5
EINECS: 202-680-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR