Tisk…

L(+)-kyselina vinná ≥99.7% FCC, FG, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

M-Clarity™ quality level = MQ300

W304409-10KG W304409-1KG W304409-SAMPLE
SIALW304409-10KGEA 8640 CZK
SIALW304409-10KG SIALW304409-1KG SIALW304409-SAMPLE
L(+)-kyselina vinná ≥99.7% FCC, FG, Sigma-Aldrich®
L(+)-kyselina vinná
Vzorec: C₄H₆O₆
Bod varu: 399 °C (1013 hPa)
Bod tání: 168…170 °C
Hustota: 1,759 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS číslo: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR