Tisk…

Linalyl acetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,7-Dimethylocta-1,6-dien-3-yl acetate

M-Clarity™ quality level = MQ300

W263613-100G-K W263613-1KG-K
SIALW263613-100G-KEA 3340 CZK
SIALW263613-100G-K SIALW263613-1KG-K
Linalyl acetate, Sigma-Aldrich®
Linalyl acetate
Vzorec: C₁₂H₂₀O₂
MDL Number: MFCD00008907
CAS číslo: 115-95-7
EINECS: 204-116-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR