Tisk…
Dehydrated culture media, WL nutrient broth
Dodavatel: Merck
W2261-500G
SIALW2261-500GEA 0 CZK
SIALW2261-500G
Dehydrated culture media, WL nutrient broth
Média mikrobiologická Mikrobiologická média dehydratovaná
WL nutrient broth consists of casein enzymatic hydrolysate (5,00 g/l), yeast extract (4,00 g/l), dextrose (50,00 g/l), monopotassium phosphate (0.55 g/l), potassium chloride (0.425 g/l), calcium chloride (0.125 g/l), manganese sulphate (0.125 g/l), ferric chloride (0.0025 g/l), manganese sulphate (0.0025 g/l) and bromocresol green (0.022 g/l).
Used for the cultivation of bacteria encountered in breweries and industrial fermentations.