Tisk…

Thymidine (2'-deoxythymidine), Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

T9250-1G T9250-5G T9250-10G T9250-25G
SIALT9250-1GEA 1760 CZK
SIALT9250-1G SIALT9250-5G SIALT9250-10G SIALT9250-25G
Thymidine (2'-deoxythymidine), Sigma-Aldrich®
Thymidine (2'-deoxythymidine)
Vzorec: C₁₀H₁₄N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00006537
CAS číslo: 50-89-5
EINECS: 200-070-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR