Tisk…

Tetramethylammonium hydroxide pentahydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

T7505-100G T7505-25G T7505-5G
SIALT7505-100GEA 8920 CZK
SIALT7505-100G SIALT7505-25G SIALT7505-5G
Tetramethylammonium hydroxide pentahydrate, Sigma-Aldrich®
Tetramethylammonium hydroxide pentahydrate
Vzorec: C₄H₁₃NO·5H₂O
MW: 181.23
Bod tání: 65…68 °C
Hustota: 1.829
MDL Number: MFCD00149566
CAS číslo: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR