Tisk…

Tetracycline hydrochloride, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

T3383-25G T3383-100G
SIALT3383-25GEA 2350 CZK
SIALT3383-25G SIALT3383-100G
Tetracycline hydrochloride, Sigma-Aldrich®
Tetracycline hydrochloride
Vzorec: C₂₂H₂₅ClN₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS číslo: 64-75-5
EINECS: 200-593-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR