Tisk…

Tricin, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ400

T0377-100G T0377-1KG T0377-250G T0377-25G T0377-25KG T0377-5KG
SIALT0377-100GEA 6480 CZK
SIALT0377-100G SIALT0377-1KG SIALT0377-250G SIALT0377-25G SIALT0377-25KG SIALT0377-5KG
Tricin, Sigma-Aldrich®
Tricin
Vzorec: (HOCH₂)₃CNHCH₂CO₂H
Bod tání: 187 °C
Hustota: 1,523 g/cm³ (19.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS číslo: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR