Tisk…

Starch (from corn), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

S4126-2KG S4126-5KG
SIALS4126-2KGEA 1950 CZK
SIALS4126-2KG SIALS4126-5KG
Starch (from corn), Sigma-Aldrich®
Starch
Vzorec: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS číslo: 9005-25-8
EINECS: 232-679-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR