Tisk…

1,4-Diaminobutane dihydrochloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ400

P7505-100G P7505-25G
SIALP7505-100GEA 8250 CZK
SIALP7505-100G SIALP7505-25G
1,4-Diaminobutane dihydrochloride, Sigma-Aldrich®
1,4-Diaminobutane dihydrochloride
Vzorec: C₄H₁₄Cl₂N₂
Hustota: 1,31 g/cm³ (20 °C)
MDL Number: MFCD00012526
CAS číslo: 333-93-7
EINECS: 206-375-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR