Tisk…
Poly-L-lysine hydrobromide
Dodavatel: Merck
P6282-5MG
SIALP6282-5MGEA 3280 CZK
SIALP6282-5MG
Poly-L-lysine hydrobromide
Proteiny
Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water.
  • Suitable for cell culture
Recommended as a cell culture substratum when using 0.5 ml of a 0.1 mg/ml solution to coat 25 cm2. Optimal conditions for attachment must be determined for each cell line and application.
Upozornení: Sterile solutions are stable for up to 2 years when stored at 2 to 8 °C. It should be stored desiccated at –20 °C.