Tisk…

Propagyl bromide ∼9.2 mol/l in toluene ≥80%, Sigma-Aldrich®

Dodavatel: Merck

P51001-5KG P51001-125G P51001-50G P51001-625G
SIALP51001-5KGEA 71020 CZK
SIALP51001-5KG SIALP51001-125G SIALP51001-50G SIALP51001-625G
Propagyl bromide ∼9.2 mol/l in toluene ≥80%, Sigma-Aldrich®
Propagyl bromide
Vzorec: C₃H₃Br
MDL Number: MFCD00000241
CAS číslo: 106-96-7
EINECS: 203-447-1
UN: 2924
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR