Tisk…

Propargyl amine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Monopropagyl amine, Prop-2-ynylamine, 3-Aminopropyne, Prop-2-yn-1-amine

M-Clarity™ quality level = MQ200

P50900-5G P50900-1G P50900-25G
SIALP50900-5GEA 2620 CZK
SIALP50900-5G SIALP50900-1G SIALP50900-25G
Propargyl amine, Sigma-Aldrich®
Propargyl amine
Vzorec: C₃H₅N
Bod varu: 83 °C (1013 hPa)
Hustota: 0,867 g/cm³ (16 °C)
MDL Number: MFCD00008198
CAS číslo: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR