Tisk…

(±)-Pipecolinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (±)-2-Piperidinecarboxylic acid, DL-Pipecolinic acid, H-DL-Homopro-OH, Pipecolinic Acid, Piperidine-2-carboxylic acid, Pipecolinic acid

M-Clarity™ quality level = MQ100

P45850-100G P45850-25G
SIALP45850-100GEA 9140 CZK
SIALP45850-100G SIALP45850-25G
(±)-Pipecolinic acid, Sigma-Aldrich®
(±)-Pipecolinic acid
Vzorec: C₆H₁₁NO₂
MW: 129.16
MDL Number: MFCD00064347
CAS číslo: 535-75-1
EINECS: 208-616-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR