Tisk…

Penicillin/Streptomycin/Neomycin solution_SC (~5,000 units penicillin, 5 mg streptomycin and 10 mg neomycin/ml) Stabilised, bioreagent Cell Culture, Filter sterile

Dodavatel: Merck
Nebezpečí

Synonyms: Pen-Strep-Neo

MQ200

P4083-100ML P4083-20ML
SIALP4083-100MLEA 2220 CZK
SIALP4083-100ML SIALP4083-20ML
Penicillin/Streptomycin/Neomycin solution_SC (~5,000 units penicillin, 5 mg streptomycin and 10 mg neomycin/ml) Stabilised, bioreagent Cell Culture, Filter sterile
Penicillin/Streptomycin/Neomycin

Used as a cell culture media supplement to control bacterial contamination


Mode of Action: This product acts in three ways. Penicillin inhibits bacterial cell wall synthesis. Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome, causing miscoding. Neomycin inhibits initiation and elongation during protein synthesis.

Objednat

Výsledky specifikačního testu

Appearance (Form) Liquid
Appearance (Turbidity) Clear
Sterility Pass
Endotoxin Level at 10 ml/L < 0.5 EU/ml
Cell Line Hep-2 Human Cells
Cell Culture Test (percent of control) > 50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR