Tisk…

Polymyxin B sulphate, powder USP, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

P0972-10MU P0972-1MU P0972-50MU
SIALP0972-10MUEA 17450 CZK
SIALP0972-10MU SIALP0972-1MU SIALP0972-50MU
Polymyxin B sulphate, powder USP, Sigma-Aldrich®
Polymyxin B sulphate
Vzorec: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
MW: 1385.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00131991
CAS číslo: 1405-20-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR