Tisk…

Okadaic acid ammonium salt, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

O8010-10UG O8010-25UG O8010-50UG
SIALO8010-10UGEA 4390 CZK
SIALO8010-10UG SIALO8010-25UG SIALO8010-50UG
Okadaic acid ammonium salt, Sigma-Aldrich®
Okadaic acid ammonium salt
Vzorec: C₄₄H₆₇O₁₃·NH₄
MDL Number: MFCD00153856
CAS číslo: 155716-06-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR