Tisk…

Octyl-β-D-thioglucopyranoside, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

O6004-10G O6004-1G O6004-5G
SIALO6004-10GEA 37560 CZK
SIALO6004-10G SIALO6004-1G SIALO6004-5G
Octyl-β-D-thioglucopyranoside, Sigma-Aldrich®
Octyl-β-D-thioglucopyranoside
Vzorec: C₁₄H₂₈O₅S
Storage Temperature: Freezer
MDL Number: MFCD00012189
CAS číslo: 85618-21-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR