Tisk…

(4-Nitrophenyl)-β-D-galactopyranoside, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

N1252-10G N1252-1G N1252-250MG N1252-500MG N1252-5G
SIALN1252-10GEA 68380 CZK
SIALN1252-10G SIALN1252-1G SIALN1252-250MG SIALN1252-500MG SIALN1252-5G
(4-Nitrophenyl)-β-D-galactopyranoside, Sigma-Aldrich®
(4-Nitrophenyl)-β-D-galactopyranoside
Vzorec: C₁₂H₁₅NO₈
Storage Temperature: Freezer
MDL Number: MFCD00063256
CAS číslo: 3150-24-1
EINECS: 221-584-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR